Order Entry
Italy
ContactUsLinkComponent
(2S,3S,5R,6R)-5,6-Bis(azidomethyl)-2,3-dimethoxy-2,3-dimethyl-1,4-dioxane ≥97.0% (by GC)
Flammable
Irritant
(2S,3S,5R,6R)-5,6-Bis(azidomethyl)-2,3-dimethoxy-2,3-dimethyl-1,4-dioxane ≥97.0% (by GC)
Catalog # TCIAB3694-100MG
Supplier:  TCI
CAS Number:  
Flammable
Irritant
(2S,3S,5R,6R)-5,6-Bis(azidomethyl)-2,3-dimethoxy-2,3-dimethyl-1,4-dioxane ≥97.0% (by GC)
Catalog # TCIAB3694-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Melting point:
    80.0 to 84.0 deg-C
  • Specific rotation [a]20/D:
    +100 to +104 deg (C=2, CHCl3)

Specification Test Results

About this item

 :
 :286.29 g/mol
UN:1325
ADR:4.1,III
 :GHS02
 :GHS07
CAS Number:1585236-34-5
Formula:C₁₀H₁₈N₆O₄