
2-Bromothieno[2,3-b]thiophene ≥98.0% (by GC)
Catalog # TCIAB3692-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:36.0 to 40.0 deg-C
Specification Test Results
About this item
:
:219.11 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:25121-81-7
Formula:C₆H₃BrS₂