
4-tert-Butyl-2,6-dimethylphenylsulphur trifluoride ≥90.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:947725-04-4
:
Formula:C₁₂H₁₇F₃S
MDL Number:MFCD11858620
:250.32 g/mol
Storage Temperature:Refrigerator
UN:3261
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White to Light yellow to Light red powder to crystal
- Purity (Neutralization titration):min. 90.0 %
Specification Test Results
Frequently Bought Together