Order Entry
Italy
ContactUsLinkComponent
2-(2-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborine ≥98.0% (by GC, total nitrogen)
2-(2-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborine ≥98.0% (by GC, total nitrogen)
Supplier:  TCI
Certificates
2-(2-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborine ≥98.0% (by GC, total nitrogen)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (with Total Nitrogen):
    min. 98.0 %

Specification Test Results

Related Information

Frequently Bought Together