
tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate ≥98.0% (by HPLC, titration analysis)
: TCI
:152120-54-2
:MFCD02683427
:C₁₄H₂₂N₄O₄
:MFCD01075122
:310.354 g/mol
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:88.0 to 92.0 deg-C
Frequently Bought Together