Order Entry
Italy
ContactUsLinkComponent
 
tert-Butyl-2,6-dichloroisonicotinate ≥98.0% (by GC)
  TCIAB3595-5G
 :  TCI
 :  
undefined
tert-Butyl-2,6-dichloroisonicotinate ≥98.0% (by GC)
  TCIAB3595-5G
 :  TCI
 :  

 

  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    80.0 to 84.0 deg-C

 

 

 :
 :248.10 g/mol
 :75308-46-2
 :C₁₀H₁₁Cl₂NO₂
 :MFCD08445655