Order Entry
Italy
ContactUsLinkComponent
 
2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-di-n-octylfluorene ≥98.0% (by HPLC)
Catalog # TCIAB3582-1G
Supplier:  TCI
CAS Number:  
2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-di-n-octylfluorene ≥98.0% (by HPLC)
Catalog # TCIAB3582-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Melting point:
    127.0 to 131.0 deg-C

Specification Test Results

About this item

 :
 :642.58 g/mol
CAS Number:196207-58-6
Formula:C₄₁H₆₄B₂O₄
MDL Number:MFCD16294554