Order Entry
Italy
ContactUsLinkComponent
1,4-Bis(10-undecenyloxy)benzene ≥97.0% (by HPLC)
1,4-Bis(10-undecenyloxy)benzene ≥97.0% (by HPLC)
Catalog # TCIAB3575-1G
Supplier:  TCI
CAS Number:  
1,4-Bis(10-undecenyloxy)benzene ≥97.0% (by HPLC)
Catalog # TCIAB3575-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystaline
  • Purity (HPLC):
    min. 97.0 area%
  • Melting point:
    59.0 to 63.0 deg-C

Specification Test Results

About this item

 :
 :414.67 g/mol
Boiling Pt:160°C
CAS Number:138551-10-7
Formula:C₂₈H₄₆O₂
MDL Number:MFCD18207714