4,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,1,3-benzothiadiazole ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:934365-16-9
:
Formula:C₁₈H₂₆B₂N₂O₄S
MDL Number:MFCD11656086
:388.10 g/mol
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 95.0 %
- Melting point:212.0 to 216.0 deg-C
Specification Test Results
Related Information
Frequently Bought Together