
2,7-Dibromo-9,9-bis(6-bromohexyl)fluorene ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:570414-33-4
:
Formula:C₂₅H₃₀Br₄
MDL Number:MFCD09260450
:650.13 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 95.0 %
- Melting point:70.0 to 74.0 deg-C
Specification Test Results
Frequently Bought Together