
5-Bromo-2-formylpyridine ≥98.0% (by GC)
Catalog # TCIAB3489-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:94.0 to 98.0 deg-C
Specification Test Results
About this item
:
:186.01 g/mol
Boiling Pt:70°C
:GHS07
Storage Temperature:Refrigerator
CAS Number:31181-90-5
Formula:C₆H₄BrNO
MDL Number:MFCD04112535