Benzo[1,2-b:4,5-b']dithiophene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:267-65-2
:
Formula:C₁₀H₆S₂
:190.28 g/mol
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:196.0 to 200.0 deg-C
Specification Test Results
Frequently Bought Together