1,3-Bis[3,5-bis(trifluoromethyl)phenyl]thiourea ≥98.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:1060-92-0
:
Formula:C₁₇H₈F₁₂N₂S
MDL Number:MFCD00829878
Molecular Weight:500.31 g/mol
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 93.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:150.0 to 157.0 deg-C
Specification Test Results
Frequently Bought Together