Order Entry
Italy
ContactUsLinkComponent
trans-4-(tert-Butyldiphenylsilyloxy)-L-proline ≥98.0% (by HPLC, titration analysis)
trans-4-(tert-Butyldiphenylsilyloxy)-L-proline ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
trans-4-(tert-Butyldiphenylsilyloxy)-L-proline ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Specific rotation [a]20/D:
    -35.0 to -34.0 deg (C=0.8, MeOH)

Specification Test Results

Related Information

Frequently Bought Together