
1,4-Butanediolbis(thioglycolate) ≥95.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:10193-95-0
:
Formula:C₈H₁₄O₄S₂
MDL Number:MFCD00266655
:238.32 g/mol
EINECS:233-479-1
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %
- Purity (Iodometric Titration):min. 95.0 %
Specification Test Results
Frequently Bought Together