Order Entry
Italy
ContactUsLinkComponent
(E)-5-(2-Bromovinyl)-2'-deoxyuridine ≥98.0% (by HPLC, titration analysis)
(E)-5-(2-Bromovinyl)-2'-deoxyuridine ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB3404-100MG
Supplier:  TCI
CAS Number:  
(E)-5-(2-Bromovinyl)-2'-deoxyuridine ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB3404-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %
  • Specific rotation [a]20/D:
    +8.0 to +10.0 deg (C=1, MeOH)

Specification Test Results

About this item

 :
 :333.14 g/mol
Merck Index:14,01378
Storage Temperature:Refrigerator
CAS Number:69304-47-8
Formula:C₁₁H₁₃BrN₂O₅
MDL Number:MFCD00058585