trans-2-Butene-1,4-diol ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:821-11-4
:
Formula:C₄H₈O₂
MDL Number:MFCD00063207
:88.11 g/mol
Boiling Pt:127°C
Melting Pt:25°C
:GHS07
Specification Test Results
- Appearance:White or Colorless to Yellow powder to lump to clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together