Order Entry
Italy
ContactUsLinkComponent
5,5'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,2'-bithiophene ≥98.0% (by titrimetric analysis)
5,5'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,2'-bithiophene ≥98.0% (by titrimetric analysis)
Catalog # TCIAB3363-5G
Supplier:  TCI
CAS Number:  
5,5'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,2'-bithiophene ≥98.0% (by titrimetric analysis)
Catalog # TCIAB3363-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Gray to Brown powder to crystal
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    204.0 to 210.0 deg-C

Specification Test Results

About this item

 :
 :418.18 g/mol
CAS Number:239075-02-6
Formula:C₂₀H₂₈B₂O₄S₂
MDL Number:MFCD06798112