Order Entry
Italy
ContactUsLinkComponent
1,2-Bis(phenylsulphinyl)ethane ≥97.0% (by HPLC)
1,2-Bis(phenylsulphinyl)ethane ≥97.0% (by HPLC)
Catalog # TCIAB3352-5G
Supplier:  TCI
CAS Number:  
1,2-Bis(phenylsulphinyl)ethane ≥97.0% (by HPLC)
Catalog # TCIAB3352-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 97.0 % (total of isomer)

Specification Test Results

About this item

 :
 :278.38 g/mol
CAS Number:6099-21-4
Formula:C₁₄H₁₄O₂S₂
MDL Number:MFCD11846120