Order Entry
Italy
ContactUsLinkComponent
4-Bromomethyl-1,2-dihydroquinoline-2-one ≥97.0% (by HPLC, total nitrogen)
Irritant
4-Bromomethyl-1,2-dihydroquinoline-2-one ≥97.0% (by HPLC, total nitrogen)
Supplier:  TCI
Certificates
Irritant
4-Bromomethyl-1,2-dihydroquinoline-2-one ≥97.0% (by HPLC, total nitrogen)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (with Total Nitrogen):
    min. 97.0 %

Specification Test Results

Related Information

Frequently Bought Together