Order Entry
Italy
ContactUsLinkComponent
4-(tert-Butoxycarbonylaminomethyl)cyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
4-(tert-Butoxycarbonylaminomethyl)cyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
Catalog # TCIAB3251-5G
Supplier:  TCI
CAS Number:  
4-(tert-Butoxycarbonylaminomethyl)cyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
Catalog # TCIAB3251-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 % (total of isomers)
  • Purity (Neutralization titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :257.33 g/mol
CAS Number:162046-58-4
Formula:C₁₃H₂₃NO₄