β-methasone-21-acetate ≥98.0% (by HPLC)
: TCI
:987-24-6
:
:C₂₄H₃₁FO₆
:MFCD00867462
:434.50 g/mol
:14,01180
:213-578-6
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:213.0 to 217.0 deg-C
Frequently Bought Together