N-ɑ-Fmoc-N-ε-boc-L-lysine ≥98.0% (by HPLC, titration analysis)
: TCI
: N-ε-(tert-Butoxycarbonyl)-N-ɑ-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-lysine, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-((tert-butoxycarbonyl)amino)hexanoic acid, N-ε-Boc-N-ɑ-Fmoc-L-lysine, Fmoc-Lys(Boc)-OH
:71989-26-9
:
:C₂₆H₃₂N₂O₆
:MFCD00037138
:468.55 g/mol
:124°C
:Refrigerator
:276-256-4
- Environmentally Preferable:
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Specific rotation [a]20/D:-10.0 to -14.0 deg (C=1, DMF)
Frequently Bought Together