
(S,S)-1,2-Bis[(2-methoxyphenyl)phenylphosphino]ethane ≥97.0% (by GC, titration analysis)
TCIAB3036-100MG
: TCI
:
- Pk:100 mg
- Seq:2440
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 97.0 %
- Optical purity (LC):min. 98.0 ee%
- Purity (Iodometric Titration):min. 97.0 %
- Melting point:102.0 to 106.0 deg-C
:
:458.48 g/mol
:GHS07
:97858-62-3
:C₂₈H₂₈O₂P₂
:MFCD05863547