
3-(Bromoacetyl)pyridine ydrobromide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:17694-68-7
:
Formula:C₇H₆BrNO
MDL Number:MFCD00052182
:280.95 g/mol
EINECS:241-692-6
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together