Order Entry
Italy
ContactUsLinkComponent
9-(2-Biphenylyl)-10-methylacridinium perchlorate ≥98.0% (by HPLC, total nitrogen)
Oxidizing
Irritant
9-(2-Biphenylyl)-10-methylacridinium perchlorate ≥98.0% (by HPLC, total nitrogen)
Supplier:  TCI
Certificates
undefined
Oxidizing
Irritant
9-(2-Biphenylyl)-10-methylacridinium perchlorate ≥98.0% (by HPLC, total nitrogen)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %

Specification Test Results

Related Information

Frequently Bought Together