
10-Bromodec-1-ene ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:62871-09-4
:
Formula:C₁₀H₁₉Br
MDL Number:MFCD00078660
:219.1600 g/mol
Boiling Pt:150°C
Flash Pt:96°C
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together