4-sec-Butyl-2,6-di-tert-butylphenol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:17540-75-9
:
Formula:C₁₈H₃₀O
MDL Number:MFCD00075575
:262.44 g/mol
Boiling Pt:141°C
EINECS:241-533-0
:GHS07
:GHS08
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together