Order Entry
Italy
ContactUsLinkComponent
 
 
Bromine-1,4-dioxane complex ≥95.0% (by titrimetric analysis)
 :  TCI
 
 
Bromine-1,4-dioxane complex ≥95.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (Iodometric Titration):
    min. 95.0 %
  • Melting point:
    66.0 to 70.0 deg-C

 

Frequently Bought Together