3-Bromo-1,10-phenanthroline ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB2632-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:171.0 to 175.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:259.11 g/mol
CAS Number:66127-01-3
Formula:C₁₂H₇BrN₂
MDL Number:MFCD08276287