
3-Bromo-4-formylpyridine ≥98.0% (by GC)
: TCI
:70201-43-3
:
:C₆H₄BrNO
:MFCD05864506
:186.01 g/mol
:GHS07
- Appearance:White to Yellow to Orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:78.0 to 82.0 deg-C
Frequently Bought Together