Order Entry
Italy
ContactUsLinkComponent
4-(4-Bromophenyl)thiomorpholine-1,1-dioxide ≥98.0% (by GC, titration analysis)
4-(4-Bromophenyl)thiomorpholine-1,1-dioxide ≥98.0% (by GC, titration analysis)
Catalog # TCIAB2614-5G
Supplier:  TCI
CAS Number:  
4-(4-Bromophenyl)thiomorpholine-1,1-dioxide ≥98.0% (by GC, titration analysis)
Catalog # TCIAB2614-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    190.0 to 194.0 deg-C
  • Solubility in hot Toluene:
    almost transparency

Specification Test Results

About this item

 :
 :290.18 g/mol
CAS Number:1093878-42-2
Formula:C₁₀H₁₂BrNO₂S