
Bis(trifluoromethanesulphonyl)imide ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:82113-65-3
:
Formula:C₂HF₆NO₄S₂
MDL Number:MFCD00214154
:281.143 g/mol
Boiling Pt:92°C
Storage Temperature:Refrigerator
UN:3261
ADR:8,I
:GHS05
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 99.0 %
Specification Test Results
Frequently Bought Together