4,4'-Bicyclohexanone ≥98.0% (by GC)
Catalog # TCIAB2405-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:114.0 to 118.0 deg-C
Specification Test Results
About this item
:
:194.27 g/mol
CAS Number:23391-99-3
Formula:C₁₂H₁₈O₂
MDL Number:MFCD00210694