N-(4-Benzyloxytrityl)-N'-(2-nitrobenzenesulphonyl)-1,3-diaminopropane resin 0.9 - 1.1 mmol/g, cross-linked with 1% DVB (200-400 mesh)
Catalog # TCIAB2371-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Loading:0.9 to 1.1 mmol/g
Specification Test Results
About this item
: