

(1S,2S)-N,N'-Bis(2-acetyl-3-oxo-2-butenylidene)-1,2-dimesitylethylenediaminato cobalt(II)
TCIAB2315-100MG
: TCI
:
- Pk:100 mg
- Seq:2440
- Appearance:Orange to Brown powder to crystal
- Cobalt:min. 10.0 %
- Functionality test:to pass test
:
:573.60 g/mol
:GHS07
:GHS08
:259259-80-8
:C₃₂H₃₈CoN₂O₄
:MFCD06797062