Order Entry
Italy
ContactUsLinkComponent
(1R,2R)-N,N'-Bis(2-acetyl-3-oxo-2-butenylidene)-1,2-dimesitylethylenediaminato cobalt(II)
Irritant
Health
(1R,2R)-N,N'-Bis(2-acetyl-3-oxo-2-butenylidene)-1,2-dimesitylethylenediaminato cobalt(II)
Catalog # TCIAB2314-100MG
Supplier:  TCI
CAS Number:  
Irritant
Health
(1R,2R)-N,N'-Bis(2-acetyl-3-oxo-2-butenylidene)-1,2-dimesitylethylenediaminato cobalt(II)
Catalog # TCIAB2314-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown powder to crystal
  • Cobalt:
    min. 10.0 %

Specification Test Results

About this item

 :
 :573.60 g/mol
 :GHS07
 :GHS08
CAS Number:361346-80-7
Formula:C₃₂H₃₈CoN₂O₄
MDL Number:MFCD06797061