1,4-Bis(2-hydroxyethyl)benzene ≥98.0% (by GC)
Catalog # TCIAB2295-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:86.0 to 90.0 deg-C
Specification Test Results
About this item
:
:166.22 g/mol
Boiling Pt:193°C
Melting Pt:88°C
CAS Number:5140-03-4
Formula:C₁₀H₁₄O₂
MDL Number:MFCD00666548