Order Entry
Italy
ContactUsLinkComponent
N-ɑ-Boc-N(in)-formyl-L-tryptophan ≥98.0% (by titrimetric analysis)
N-ɑ-Boc-N(in)-formyl-L-tryptophan ≥98.0% (by titrimetric analysis)
Catalog # TCIAB2260-5G
Supplier:  TCI
CAS Number:  
N-ɑ-Boc-N(in)-formyl-L-tryptophan ≥98.0% (by titrimetric analysis)
Catalog # TCIAB2260-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Light gray to Light yellow powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %
  • Specific rotation [a]20/D:
    +17.0 to +25.0 deg (C=2, EtOH)
  • Solubility in Methanol:
    very faint turbidity

Specification Test Results

About this item

 :
 :332.36 g/mol
Storage Temperature:Refrigerator
CAS Number:47355-10-2
Formula:C₁₇H₂₀N₂O₅
MDL Number:MFCD00065992