
3,5-Bis[3,5-bis(3,5-dimethoxybenzyloxy)benzyloxy]benzyl bromide ≥97.0% (by titrimetric analysis)
TCIAB2248-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 96.0 area%
- Purity (Argentmetric Titration):min. 97.0 %
:
:1047.99 g/mol
:GHS07
:Refrigerator
:152811-37-5
:C₅₇H₅₉BrO₁₄