Ethyl-11-bromoundecanoate ≥96.0% (by GC)
Catalog # TCIAB2242-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
About this item
:
:293.25 g/mol
CAS Number:6271-23-4
Formula:C₁₃H₂₅BrO₂
MDL Number:MFCD00045048