Order Entry
Italy
ContactUsLinkComponent
(R)-(-)-1,1'-Binaphthyl-2,2'-diyl bis(trifluoromethanesulphonate) ≥96.0% (by HPLC)
Corrosive
(R)-(-)-1,1'-Binaphthyl-2,2'-diyl bis(trifluoromethanesulphonate) ≥96.0% (by HPLC)
Catalog # TCIAB2230-1G
Supplier:  TCI
CAS Number:  
Corrosive
(R)-(-)-1,1'-Binaphthyl-2,2'-diyl bis(trifluoromethanesulphonate) ≥96.0% (by HPLC)
Catalog # TCIAB2230-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystaline
  • Purity (HPLC):
    min. 96.0 area%
  • Melting point:
    82.0 to 87.0 deg-C
  • Specific rotation [a]20/D:
    -135 to -154 deg (C=1,THF)
  • Lambda max.:
    215.0 to 225.0 nm (CH3CN)

Specification Test Results

About this item

 :
 :550.44 g/mol
UN:3261
ADR:8,II
 :GHS05
CAS Number:126613-06-7
Formula:C₂₂H₁₂F₆O₆S₂
MDL Number:MFCD00274615