Bromopolystyrene resin 2.6 - 2.8 mmol/g, cross-linked with 2% DVB (200-400 mesh)
Catalog # TCIAB2172-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
Specification Test Results
About this item
:
CAS Number:88497-56-7
MDL Number:MFCD00146196