Tetrasodium-N,N-bis(carboxymethyl)-L-glutamate ∼40% in water
Supplier: TCI
Certificates
About this item
CAS Number:51981-21-6
:
Formula:C₉H₁₃NO₈
MDL Number:MFCD01862262
:351.13 g/mol
EINECS:257-573-7
Specification Test Results
- Appearance:Light orange to Yellow to Green clear liquid
- Concentration (Chelometric Method):40.0 to 44.0 w/w%
- Specific rotation [a]20/D:+7.0 to +11.0 deg (neat)
Specification Test Results
Frequently Bought Together