Order Entry
Italy
ContactUsLinkComponent
 
 
 
Boron trifluoride dibutyl etherate (∼30% BF₃)
 :  TCI
 
 
 
Boron trifluoride dibutyl etherate (∼30% BF₃)
 :  TCI

 

  • Appearance:
    Colorless to Brown clear liquid
  • Concentration (Gravimetric Method):
    29.0 to 33.0 w/w%

 

Frequently Bought Together