Order Entry
Italy
ContactUsLinkComponent
Bis(triphenylphosphine)palladium(II) diacetate ≥98.0% (by titrimetric analysis)
Bis(triphenylphosphine)palladium(II) diacetate ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
Bis(triphenylphosphine)palladium(II) diacetate ≥98.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light orange to Yellow to Green powder to crystal
  • Purity (Chelometric Titration):
    min. 98.0 %

Specification Test Results

Frequently Bought Together