
1,6-Bis(diphenylphosphino)hexane ≥95.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:19845-69-3
:
Formula:C₃₀H₃₂P₂
MDL Number:MFCD00003053
:454.53 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 95.0 %
- Purity (Iodometric Titration):min. 95.0 %
- Melting point:124.0 to 128.0 deg-C
Specification Test Results
Related Information
Frequently Bought Together