Order Entry
Italy
ContactUsLinkComponent
N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine ≥98.0% (by titrimetric analysis)
Irritant
Health
N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
Irritant
Health
N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine ≥98.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

Frequently Bought Together