Order Entry
Italy
ContactUsLinkComponent
 
1,2-Bis(4-methylphenyl)-1,2-diphenyl-1,2-ethanediol ≥96.0% (by GC)
Catalog # TCIAB1849-1G
Supplier:  TCI
CAS Number:  
1,2-Bis(4-methylphenyl)-1,2-diphenyl-1,2-ethanediol ≥96.0% (by GC)
Catalog # TCIAB1849-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 96.0 %
  • Melting point:
    155.0 to 158.0 deg-C
  • Solubility in Methanol:
    very faint turbidity

Specification Test Results

About this item

 :
 :394.51 g/mol
CAS Number:808-12-8
Formula:C₂₈H₂₆O₂
MDL Number:MFCD00026012