Order Entry
Italy
ContactUsLinkComponent
(S)-(-)-3-tert-Butoxycarbonyl-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine ≥96.0% (by GC)
Irritant
(S)-(-)-3-tert-Butoxycarbonyl-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine ≥96.0% (by GC)
Supplier:  TCI
Certificates
Irritant
(S)-(-)-3-tert-Butoxycarbonyl-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine ≥96.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 96.0 %
  • Specific rotation [a]20/D:
    -50.0 to -65.0 deg (C=2,chloroform)

Specification Test Results

Related Information

Frequently Bought Together